Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Disc Amikacin2, AN-30
Description
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents.
- Employs Bauer-Kirby disc diffusion testing method
- Use in self-tamping dispensers
- Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
- Cartridges of 50 discs each are sold individually or in packs of 10
Specifications
Specifications
| Agenti antimicrobici | Amikacin |
| Concentrazione | 30 μg |
| Tipo di imballaggio | Disc |
| Quantità | 10/Pk. |