Learn More
BD Bromodeoxyuridine (BrdU)
Product Code. 10222843 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10222843 25 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10222843 Supplier BD Supplier No. 550891
  • ONLINE EXCLUSIVE PRICE
    314.00€
    266.86€ / pezzi 1
    Risparmia 47.14€
    15% Sconto
 Restricted Item
Spedizione stimata: 19-11-2026
A thymidine analog used in cell proliferation studies
BrdU in culture is incorporated into the DNA during DNA synthesis. Cellular incorporation of BrdU can be detected by anti-BrdU specific antibodies following membrane permeabilization through flow cytometry or immunohistochemistry.
  • Free of preservatives; handle under aseptic conditions
  • Provided as five vials, each containing 0.5mL of a 10mg/mL solution diluted in 1X DPBS
TRUSTED_SUSTAINABILITY
Specifications
Format Liquid
pH 7.2
Product Type Bromodeoxyuridine (BrdU)
Content And Storage -80°C
Concentration 10 mg/mL
Quantity 25 mg
For Use With (Application) Cellular incorporation of BrdU can be detected by anti-BrdU specific Antibodies following membrane permeabilization through flow cytometry or immunohistochemistry