Learn More
BD CBA Mouse Th1/Th2 Cytokine Kit
Product Code. 15846598 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
80 Tests
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15846598 80 Tests 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15846598 Supplier BD Supplier No. 551287
  • ONLINE EXCLUSIVE PRICE
    1930.00€
    1642.12€ / 1 sets
    Risparmia 287.88€
    14% Sconto
 Restricted Item
Spedizione stimata: 18-11-2026
Includes
Reagents for capture and detection of IL-2, IL-4, IL-5, IFN-γ, and TNF; standards; wash buffer; assay diluent; cytometer setup reagents
Optimized for analysis of physiologically relevant concentrations (pg/mL levels) of specific cytokine proteins in tissue culture supernatants and serum samples
BD CBA assays provide a method of capturing a soluble analyte or set of analytes with beads of known size and fluorescence, making it possible to detect analytes using flow cytometry.
  • Can be used to measure Interleukin-2 (IL-2), Interleukin-4 (IL-4), Interleukin-5 (IL-5), Interferon-γ (IFN-γ), and Tumor Necrosis Factor (TNF) protein levels in a single sample
  • Provides sufficient reagents for analysis of 80 samples
TRUSTED_SUSTAINABILITY
Specifications
Detects IL-2, IL-4, IL-5, IFN-g and TNF
For Use With (Application) For analysis of physiologically relevant concentrations (pg/mL levels) of specific Cytokine Proteins in Tissue Culture supernatants and Serum samples
Quantity 80 Tests
Content And Storage 2°C to 8°C
For research use only.