Learn More
BD Cytometric Bead Array (CBA) Human IL-10; Flex Set, Bead B7
Codice prodotto. 15809278
Click to view available options
Quantità:
100 Tests
Dimensione della confezione:
Pezzo
Codice prodotto. 15809278
Marca: BD 558274
Includes
Human IL-10; Standard; Human IL-10; PE Detection Reagent; Human IL-10; Capture Bead B7
Bead-based immunoassay capable of measuring human interleukin-10 (IL-10) in serum, plasma, and cell culture supernatant samples
Human reactivity was determined by testing samples with the BD CBA Human IL-10 Flex Set
Flow cytometry
TRUSTED_SUSTAINABILITY
Specifica
Antigene IL-10
Cross-reattività Human
Include Human IL-10, standard, Human IL-10, PE detection reagent, Human IL-10, capture bead B7
Quantità 100 Tests
Tipo di campione Serum, Plasma, Cell Culture Supernatant
Content And Storage 4°C
Da utilizzare con (applicazione) Flow Cytometry
Product Type Human IL-10 flex set (bead B7)