Learn More
Cytometric Bead Array (CBA) Mouse/Rat Soluble Protein Master Buffer Kit, 100 tests, BD
Codice prodotto. 10318813 Sfoglia Tutto BD Biosciences Prodotti
Cambia vista
Click to view available options
Quantità:
100 Tests
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
10318813 100 Tests Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 10318813 Fornitore BD Biosciences N. del fornitore 558266
  • ONLINE EXCLUSIVE PRICE
    420.00€
    357.03€ / pezzi 1
    Risparmia 62.97€
    14% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 12-11-2026
Includes
Assay Diluent, Capture Bead Diluent, Detection Reagent Diluent, Wash Buffer, Instrument Setup Beads (A1, A9, F1, F9), PE Instrument Setup Bead F1, PE Positive Control Detector
Employs particles with discrete fluorescence intensities to detect soluble analytes at very low concentrations
This kit contains all supporting reagents necessary to perform an assay using BD™ CBA Mouse or Rat Soluble Protein Flex Set. Buffers and instrument setup reagents are optimized for analysis of analytes in tissue culture supernatants, plasma and serum samples.
  • Analyte working assay range: 10 to 2500pg/mL (mouse), 40 to 10000pg/mL (rat)
  • Should not be used with any non-Mouse or Rat Soluble Protein Flex Sets
  • Available in pack sizes of 100 and 500 tests
Flow cytometry
TRUSTED_SUSTAINABILITY
Specifica
Content And Storage 2°C to 8°C
N. di test 100
Include 1 bottle 30mL assay diluent, 1 bottle 5mL capture bead diluent, 1 bottle 5mL detection reagent diluent, 1 bottle 130mL wash buffer, 1 vial 0.25mL instrument setup bead A1, 1 vial 0.25mL instrument setup bead A9, 1 vial 1.0mL instrument setup bead F1
Quantità 100 Tests