Learn More
BD Horizon™ PE-CF594 Annexin V
Product Code. 15892869 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15892869 50 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15892869 Supplier BD Supplier No. 563544
  • ONLINE EXCLUSIVE PRICE
    465.00€
    200.37€ / pezzi 1
    Risparmia 264.63€
    56% Sconto
 Restricted Item
Spedizione stimata: 04-12-2026
A sensitive probe for identifying apoptotic cells, binding to negatively charged phospholipid surfaces with a higher affinity for phosphatidylserine (PS) than most other phospholipids
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Flow Cytometry
Quantity 50 Tests
Color PE-CF594
Content And Storage Store undiluted at 4°C
Form Buffered Solution
For Research Use Only