Learn More
BD Mouse Intracellular Cytokine Staining Starter Kit
Product Code. 15839698 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 Tests
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15839698 25 Tests 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15839698 Supplier BD Supplier No. 559311
  • ONLINE EXCLUSIVE PRICE
    1000.00€
    848.85€ / 1 sets
    Risparmia 151.15€
    15% Sconto
 Restricted Item
Spedizione stimata: 03-12-2026
Includes
BD PharMingen Stain Buffer (FBS); Cytofix/Cytoperm Buffer; Perm/Wash Buffer; PE-conjugated anti-human or mouse IL-2; PE-conjugated anti-human or mouse IFN-g; PE-conjugated anti-human or mouse TNF-a; PE-conjugated isotype control cocktail; Purified blocking antibody cocktail; Leukocyte Activation Cocktail; MiCK-1 or HiCK-1 Cytokine Positive Control Cells; Staining Kit Manual
Serves as an introduction to the intracellular staining technique
  • Contains the essential reagents for the activation of a mouse spleen cell population and their subsequent intracellular staining to detect IL-2, IFN-γ-, and TNF-a-producing cells
TRUSTED_SUSTAINABILITY
Specifications
Detects IL-2, IFN-g- and TNF-a
Quantity 25 Tests
Content And Storage 4°C