Learn More
BD Organelle Sampler Kit
Codice prodotto. 15846279 Sfoglia Tutto BD Biosciences Prodotti
Cambia vista
Click to view available options
Quantità:
1 Kit
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
15846279 1 Kit Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Highly toxic, dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing
Codice prodotto. 15846279 Fornitore BD N. del fornitore 612740
  • ONLINE EXCLUSIVE PRICE
    2145.00€
    1825.19€ / pezzi 1
    Risparmia 319.81€
    14% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 19-11-2026
Highly toxic, dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing
Includes
Annexin II, BiP/GRP78, b-Catenin, Caveolin 1, Connexin-43, EEA1, GM130, Integrina2/VLA-2a, Lamp-1, MAP2B, Bcl-2, Nucleoporin p62, Paxillin, ZO-1, Storage Buffer: aqueous buffered solution containing BSA, glycerol, and ≥0.09% sodium azide
For blotting and immunoassay procedures
  • Concentration: 250μg/mL
Western Blotting, Immunoprecipitation, Immunofluorescence, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifica
Quantità 1 Kit