Learn More
BD Perm/Wash™ Buffer
Product Code. 15827868 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15827868 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15827868 Supplier BD Supplier No. 554723
  • ONLINE EXCLUSIVE PRICE
    255.00€
    216.77€ / pezzi 1
    Risparmia 38.23€
    14% Sconto
 Restricted Item
Spedizione stimata: 27-11-2026
Can be used in intracellular cytokine staining to permeabilize cells and as an antibody diluent and cell wash buffer
  • 10X stock solution containing both Fetal Bovine Serum (FBS) and saponin
It is not uncommon for the color of this product to vary from lot to lot. A small amount of insoluble precipitate is also common. Color variation and/or precipitate do not affect product performance.
TRUSTED_SUSTAINABILITY
Specifications
Formulation Fetal Bovine Serum (FBS) Solution 10X and Saponin
For Use With (Application) Can be used in intracellular Cyto kine staining to permeabilize cells and as an Antibody diluent and cell Wash Buffer
pH Range 7.2 to 7.6
Content And Storage 4°C
Product Type Perm or Wash Buffer
Quantity 100 mL
Format Liquid