Learn More
Phosflow Perm Buffer III, BD
Product Code. 15849178 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
125 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15849178 125 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15849178 Supplier BD Biosciences Supplier No. 558050
  • ONLINE EXCLUSIVE PRICE
    329.00€
    279.60€ / pezzi 1
    Risparmia 49.40€
    15% Sconto
 Restricted Item
Spedizione stimata: 07-12-2026
For the permeabilization of fixed Peripheral Blood Mononuclear Cells (PBMCs) or lysed whole blood
  • Designed for optimal fluorescent staining in flow cytometry
  • Can be used with antibodies for phospho-protein analysis
  • In many cases, may be used simultaneously with phospho-specific antibodies and the staining of cell surface markers (e.g CD markers)
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Content And Storage Room Temperature
Format Liquid
Quantity 125 mL
Validated Application RUO
Product Type Staining Buffer
compliance-icons compliance-icons compliance-icons
Product Identifier
  • perm bufferr III
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: methanol