Learn More
Bel-Art™ SP Scienceware™ Safety Cutter
Easily and safely cut LabMat™ liners, plastic “clamshell” packaging, shrink wrap, paper, etc.
Marca: Bel-Art™ F21012-0000
Dettagli aggiuntivi : Peso : 0.20000kg
Descrizione
- Faster and safer than scissors or utility knives: will not cut fingers
- Ceramic blade stays rust-free and remains sharp longer than stainless steel blades
- Blade does not need to be covered for safety
- Elongated tear-drop shape measures 2.37 x 1.18 x 0.18 in. (6 x 3 x 0.5cm)
Specifica
Ceramic | |
Safety Cutter | |
Easily cuts LabMat™ liner, clamshell packaging, Plastic bags, Shrink-wrap, Paper and much more |