Learn More
Jenway™ Benchtop Dissolved Oxygen Meter
Click to view available options
Tensione:
120 V
Dimensione della confezione:
Pezzo
Descrizione
- LCD provides clear, direct readout of percentage of dissolved oxygen, mg/L, or BOD of samples
- Easily export data to computer or printer via RS-232 or IrDA connections
- % Saturation from 0 to 199 or 0 to 25 with accuracy of ±2% with 50°F (10°C) of calibration temperature
- Manual salinity and barometric pressure compensation
- Features real-time clock
Specifica
Specifica
| Punti dati | DO, temperature |
| Lingue | English, French, German, Italian, Spanish, Portuguese |
| Consumo energetico | 9 V power supply |
| Tensione | 120 V |