Learn More
Bensulfuron, TRC
Codice prodotto. 30338265
Cambia vista
Click to view available options
Quantità:
250 mg
Dimensione della confezione:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30338265 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30338265 Fornitore Toronto Research Chemicals N. del fornitore TRCB290060250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico Bensulfuron (Free Acid)
CAS 99283-01-9
Formula molecolare C15 H16 N4 O7 S
Quantità 250 mg
Sinonimo 2-[[[[[(4,6-Dimethoxy-2-pyrimidinyl)amino]carbonyl]amino]sulfonyl]methyl]benzoic acid, Bensulfuron, Bensulfuron (Free Acid)
InChI Formula InChI=1S/C15H16N4O7S/c1-25-11-7-12(26-2)17-14(16-11)18-15(22)19-27(23,24)8-9-5-3-4-6-10(9)13(20)21/h3-7H,8H2,1-2H3,(H,20,21)(H2,16,17,18,19,22)
SMILES COc1cc(OC)nc(NC(=O)NS(=O)(=O)Cc2ccccc2C(=O)O)n1
IUPAC Name 2-[(4,6-dimethoxypyrimidin-2-yl)carbamoylsulfamoylmethyl]benzoic acid
Molecular Weight (g/mol) 396.38
Peso formulazione 396.074