Learn More
Lenz Laborglasinstrumente™ Adattatore a gomito con anello in vetro
Description
- NS socket and NS cone
- 105° angle
- Vacuum connection
- Glass olive with drain tip
- Meets DIN 12594 standards
Specifications
Specifications
| Tipo | Bent Receiver Adapter with Glass Olive |
| Materiale | Duran Glass |
| Giunto rastremato standard | NS 19/26 |