Learn More
Benzaldehyde Dimethyl Acetal, TRC
Product Code. 30330832
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30330832 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30330832 Supplier Toronto Research Chemicals Supplier No. TRCB1839001G
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Benzaldehyde Dimethyl Acetal
CAS 1125-88-8
Molecular Formula C9H12O2
Quantity 1 g
Synonym (Dimethoxymethyl)benzene, Dimethoxyphenylmethane, NSC 286137, Phenyldimethoxymethane, alpha,alpha-Dimethoxytoluene, alpha-Methoxybenzyl Methyl Ether
InChI Formula InChI=1S/C9H12O2/c1-10-9(11-2)8-6-4-3-5-7-8/h3-7,9H,1-2H3
SMILES COC(OC)c1ccccc1
IUPAC Name dimethoxymethylbenzene
Molecular Weight (g/mol) 152.19
Formula Weight 152.0837