Learn More
Benzamide, TRC
Specifica
Specifica
| Chemical Name or Material | Benzamide |
| CAS | 55-21-0 |
| Molecular Formula | C7 H7 N O |
| Quantity | 5 g |
| Synonym | Benzenecarboxamide, Benzoic acid amide, Benzoylamine, NSC 3114, Phenylamide, Phenylcarboxamide |
| InChI Formula | InChI=1S/C7H7NO/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H2,8,9) |
| SMILES | NC(=O)c1ccccc1 |
| IUPAC Name | benzamide |
| Molecular Weight (g/mol) | 121.14 |
| Formula Weight | 121.0528 |