missing translation for 'onlineSavingsMsg'
Learn More

Benzonitrile, 99+%, for spectroscopy, Thermo Scientific Chemicals

Catalog Number p-64055
Click to view available options
Quantità:
50 mL
250 mL
1 L
Imballaggio:
Glass bottle
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
100-47-0
C7H5N
103.12
MFCD00001770
JFDZBHWFFUWGJE-UHFFFAOYSA-N
phenyl cyanide, cyanobenzene, benzenenitrile, benzoic acid nitrile, benzene, cyano, benzenecarbonitrile, phenylcyanide, fenylkyanid, fenylkyanid czech, benzonitril
7505
CHEBI:27991
benzonitrile
C1=CC=C(C=C1)C#N
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
100-47-0
C7H5N
103.12
MFCD00001770
JFDZBHWFFUWGJE-UHFFFAOYSA-N
phenyl cyanide, cyanobenzene, benzenenitrile, benzoic acid nitrile, benzene, cyano, benzenecarbonitrile, phenylcyanide, fenylkyanid, fenylkyanid czech, benzonitril
7505
CHEBI:27991
benzonitrile
C1=CC=C(C=C1)C#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY
Materiale o nome chimico Benzonitrile
Nota nome 99+%
Absorbance (1 cm cell vs water), (1 cm cell vs water) at 0nm, 0.01 max. at 360nm, 0.01 max. at 400nm, 0.03 max. at 335nm, 0.15 max. at 310nm, 0.3 max. at 305nm, 1 max. at 300nm
CAS min. % 99.0
CAS max % 100.0
Punto di fusione -13.0°C
Densità 1.0100g/mL
Punti di ebollizione 188.0°C to 191.0°C
Punto d'infiammabilità 71°C
Intervallo percentuale saggio 99% min. (GC)
Imballaggio Glass bottle
Formula lineare C6H5CN
Indice di rifrazione 1.5270 to 1.5290
Quantità 50 mL
Beilstein 09, 275
Residuo dopo l'evaporazione 0.0005% max.
Indice di Merck 15, 1099
Gravità specifica 1.01
Informazioni di solubilità Solubility in water: 10g/L (100°C). Other solubilities: misicble with most common organic solvents
Viscosità 1.24 mPa.s (25°C)
Peso formulazione 103.12
Percent Purity 99+%
Grado Spectroscopy
Forma fisica Liquid
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'