Learn More
Uvex™ Bermuda Trousers 7426-Graphit
Catalog Number 17227061
Click to view available options
Size:
42
46
48
50
52
54
56
58
60
62
64
66
The uvex suXXeed collection gets everything spot on: well thought-out functionality, sporty design, exciting color contrasts, unusual material combinations and a cut that fits like a glove. Can be worn in industrial and metal processing settings as well as for leisure.
  • Material: 65% polyester, 40% cotton ca. 245 g/m², twill weave
  • This is an optimized trousers on physiological points of view
  • Contrasting colored inserts, permanent well-being and ease of movement.
  • Waistband with elastic for an optimal fit, many different pockets do offer enough storage space, sporty alignment of style, suitable for industrial laundering
  • Oeko-Tex Standard 100
  • Sizes: 42 - 66
  • Care instructions: 60°C washable, chlorinate impossible, ironing: I points, dry cleaning: P, tumble dryer: I point, suitable for industrial laundering
Size 64
Sesso Men's
Colore Gray
Tipo di chiusura Buttons; Zipper
Monouso No
Spessore appro x 0.245 g/m²