missing translation for 'onlineSavingsMsg'
Learn More

beta 2-Microglobulin Antibody (B2M/961), Novus Biologicals™

Codice prodotto. p-5539125 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
0.02 mg
0.1 mg
Dimensione della confezione:
0.02mg
0.10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18112158 0.02 mg 0.02mg
18757903 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18112158 Fornitore Novus Biologicals N. del fornitore NBP2444710.02MG

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence

beta 2-Microglobulin Monoclonal specifically detects beta 2-Microglobulin in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Cell Surface).
TRUSTED_SUSTAINABILITY

Specifica

Antigene beta 2-Microglobulin
Applicazioni Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Classificazione Monoclonal
Clone B2M/961
Concentrazione 0.2mg/mL
Coniugato Unconjugated
Diluizione Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml
Formulazione 10mM PBS and 0.05% BSA with 0.05% Sodium Azide
N. accesso geni P61769
Alias gene beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin
Simboli geni B2M
Specie ospite Mouse
Immunogeno Full length recombinant human B2M protein
Peso molecolare dell'antigene 12 kDa
Metodo di purificazione Protein A or G purified
Quantità 0.02 mg
Status giuridico RUO
Disciplina di ricerca Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology
Primario o secondario Primary
ID gene (immissione) 567
Test di specificità Recognizes a protein of 12kDa, identified as beta-2 microglobulin. Major histocompatibility complex (MHC) class 1 molecules bind to antigens for presentation on the surface of cells. The proteasome is responsible for producing these antigens from the components of foreign pathogens. MHC class 1 molecules consist of an alpha heavy chain that contains three subdomains (alpha1, alpha2, alpha3) and a non-covalent associating light chain, known as beta-2-Microglobulin. beta-2-Microglobulin associates with the alpha3 subdomain of the alpha heavy chain and forms an immunoglobulin domain-like structure that mediates proper folding and expression of MHC class 1 molecules. The alpha1 and alpha2 domains of the alpha heavy chain form the peptide antigen-binding cleft. Mutations in the beta-2-Microglobulin gene can enhance the progression of malignant melanoma phenotypes.
Target Species Human, Primate
Content And Storage Store at 4C.
Forma Purified
Isotype IgG2b κ
Vedi altri risultati Mostra meno risultati

For Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.