missing translation for 'onlineSavingsMsg'
Learn More

beta-Actin Antibody (AC-15), Novus Biologicals™

Codice prodotto. 18709583 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
0.025 mL
Dimensione della confezione:
0.10mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18709583 0.025 mL 25µL
18775843 - 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18709583 Fornitore Novus Biologicals N. del fornitore NB600501SS

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Mouse Monoclonal Antibody has been used in 442 publications

beta-Actin Monoclonal antibody specifically detects beta-Actin in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Feline, Fish, Guinea Pig, Hamster, Leech, Mammalia, Primate, Rabbit, Sheep, Squirrel, Zebrafish, Drosophila (Negative) samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), ChIP assay.
TRUSTED_SUSTAINABILITY

Specifica

Antigene beta-Actin
Applicazioni Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, ChIP Assay, Immunoassay, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), KnockDown
Classificazione Monoclonal
Clone AC-15
Coniugato Unconjugated
Diluizione Western Blot 1:5000 - 1:10000, Simple Western 1:25, Flow Cytometry, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:1000 - 1:2000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:500, Chromatin Immunoprecipitation (ChIP), Knockout Validated, KnockDown Validated
N. accesso geni P60709
Alias gene ACTB, actin, beta, actin, cytoplasmic 1, anti-beta actin, B-Actin, Beta Actin, beta actin (ac-15), beta cytoskeletal actin, Beta-actin, beta-actin ac 15, beta-actin ac15, beta-actin ac-15, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1
Simboli geni ACTB
Specie ospite Mouse
Immunogeno This beta-Actin Antibody (AC-15) was made to a slightly modified Beta-cytoplasmic actin N-terminal peptide, Ac-Asp-Asp-Asp-Ile-Ala-Ala-Leu-Val-Ile-Asp-Asn-Gly-Ser-Gly-Lys, conjugated to KLH.
Peso molecolare dell'antigene 42 kDa
Metodo di purificazione Unpurified
Quantità 0.025 mL
Status giuridico RUO
Disciplina di ricerca Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Loading Controls, Signal Transduction, Stem Cell Markers
Primario o secondario Primary
ID gene (immissione) 60
Test di specificità In staining of chicken gizzard ultrathin tissue cryosections, the antibody labels the dense bodies and longitudinal channels linking consecutive dense bodies that are also occupied by desmin and the membrane-associated dense plaque. It does not stain adult cardiac and skeletal muscles except for traces due to contaminations of the sample with non-muscle cells, or if embryonic tissue is being used. The epitope recognized by the antibody is resistant to formalin-fixed and paraffin-embedding.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Chicken, Feline, Fish, Guinea Pig, Hamster, Leech, Mammalia, Primate, Rabbit
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Forma Ascites
Isotype IgG1
Vedi altri risultati Mostra meno risultati

For Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.