Learn More
(BetaR)-Beta-[[(S)-(4-Methylphenyl)sulfinyl]amino]-4-[[tris(1-methylethyl)silyl]oxy]benzenepropanoic Acid Methyl Ester, TRC
Codice prodotto. 30469873
Cambia vista
Click to view available options
Quantità:
10 mg
Dimensione della confezione:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30469873 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30469873 Fornitore Toronto Research Chemicals N. del fornitore TRCM32101010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico (BetaR)-Beta-[((S)-(4-Methylphenyl)sulfinyl)amino]-4-[(tris(1-methylethyl)silyl)oxy]benzenepropanoic Acid Methyl Ester
CAS 943858-40-0
Formula molecolare C26H39NO4SSi
Quantità 10 mg
InChI Formula InChI=1S/C26H39NO4SSi/c1-18(2)33(19(3)4,20(5)6)31-23-13-11-22(12-14-23)25(17-26(28)30-8)27-32(29)24-15-9-21(7)10-16-24/h9-16,18-20,25,27H,17H2,1-8H3/t25-,32?/m1/s1
SMILES COC(=O)C[C@@H](NS(=O)c1ccc(C)cc1)c2ccc(O[Si](C(C)C)(C(C)C)C(C)C)cc2
Molecular Weight (g/mol) 489.74