Learn More
Tocris Bioscience™ Bexarotene
Potent and selective RXR agonist
Marca:  Tocris Bioscience™ 5819/10
Encompass
Codice prodotto. 15756775
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi »
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
Bexarotene is a potent and selective RXR agonist (EC50 values are 24, 25 and 33 nM for RXRβ, RXRγ and RXRα, respectively). Exhibits >300-fold selectivity for RXR over RAR receptors. Inhibits tumor cell invasion and migration in vitro, and inhibits metastasis and angiogenesis in mouse models. Also promotes brown adipogenic differentiation and induces brown adipogenic reprogramming from myoblasts.
Specifica
Specifica
4-[1-(5,6,7,8-Tetrahydro-3,5,5,8,8-pentamethyl-2-naphthalenyl)ethenyl]benzoic acid
153559-49-0
LGD 1069