missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals BICINE, Electrophoresis Grade, 99+%

Catalog Number p-7057642
60.26 EUR valido fino al 2024-03-29
Usa il codice promo "21615" per avere il tuo prezzo promozionale.


Click to view available options
Quantità:
25 g
100 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
150-25-4
C6H12NNaO4
185.16
MFCD00004295
MFBDBXAVPLFMNJ-UHFFFAOYSA-M
N,N-Bis(2-hydroxyethyl)glycine
8761
CHEBI:40957
2-[bis(2-hydroxyethyl)amino]acetic acid
[Na+].OCCN(CCO)CC([O-])=O
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
150-25-4
C6H12NNaO4
185.16
MFCD00004295
MFBDBXAVPLFMNJ-UHFFFAOYSA-M
N,N-Bis(2-hydroxyethyl)glycine
8761
CHEBI:40957
2-[bis(2-hydroxyethyl)amino]acetic acid
[Na+].OCCN(CCO)CC([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico BICINE
Punto di fusione ∼191°C (decomposition)
Colore White
Forma fisica Powder
Quantità 25 g
Beilstein 1769362
Indice di Merck 14,1201
Peso formulazione 163.17
Percent Purity ≥99%
Grado Electrophoresis

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'