Learn More
Bifenox, TRC
Specifica
Specifica
| Materiale o nome chimico | Bifenox |
| CAS | 42576-02-3 |
| Formula molecolare | C14 H9 Cl2 N O5 |
| Quantità | 1 g |
| Sinonimo | 2,4-Dichlorophenyl 3-(methoxycarbonyl)-4-nitrophenyl ether, 2,4-Dichlorophenyl 3'-carbomethoxy-4'-nitrophenyl ether, 2,4-Dichlorophenyl-3'-methoxycarbonyl-4'-nitrophenyl ether, 3-Carbomethoxy-2',4'-dichloro-4-nitrodiphenyl ether, Bifenox, Biphenox, Fox, Jiazhichucaomi, MC 4379, MC 79, Methyl 5-(2,4-dichlorophenoxy)-2-nitrobenzoate, Modown, Modown 4F, Weeral |
| InChI Formula | InChI=1S/C14H9Cl2NO5/c1-21-14(18)10-7-9(3-4-12(10)17(19)20)22-13-5-2-8(15)6-11(13)16/h2-7H,1H3 |
| SMILES | COC(=O)c1cc(Oc2ccc(Cl)cc2Cl)ccc1[N+](=O)[O-] |
| IUPAC Name | methyl 5-(2,4-dichlorophenoxy)-2-nitrobenzoate |
| Molecular Weight (g/mol) | 342.13 |
| Peso formulazione | 340.9858 |