Learn More
Invitrogen™ Bigfoot™ Calibration Beads
Produktkod. 17474693 Handla allt Applied Biosystems Produkter
Change view
Klicka för att se tillgängliga alternativ
Quantità:
3 x 5 mL
Förpackningsstorlek:
15mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantità Förpackningsstorlek
17474693 3 x 5 mL 15mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 17474693 Leverantör Invitrogen™ Leverantörsnummer PL00287
 Articolo soggetto a restrizioni
In stock
Lägg i varukorgen
Edge
Lägg i varukorgen
missing translation for 'validMainframeMsg'
Denna artikel kan inte returneras. Se returpolicy
A mix of three different calibration beads used in the automated QC of the Bigfoot™ Spectral Cell Sorter. Beads are retained within the Bigfoot cell sorter in a dedicated loader position and are used during the automated QC process to verify alignment and drop delay of the system.
  • A mix of three different calibration beads used in the automated QC of the Bigfoot™ Spectral Cell Sorter
  • Beads are retained within the Bigfoot cell sorter in a dedicated loader position
  • Used during the automated QC process to verify alignment and drop delay of the system
  • The onboard QC beads allow the operator to perform automated QC at any time and assist with remote support
  • The Bigfoot QC bead bottle is automatically capped to minimize evaporation from the vial
  • Loader location contains an integrated vortex to keep the beads suspended
  • Calibration protocols are fully automated
  • No pre-mixing or lot monitoring are required
  • Convenient, ready to load
TRUSTED_SUSTAINABILITY
Specifikationer
Da utilizzare con (apparecchiatura) Invitrogen™ Bigfoot™ Cell Sorter
Da utilizzare con (applicazione) Daily QC of Bigfoot Cell Sorter
Quantità 3 x 5 mL
Product Type QC Beads
Linea di prodotti Invitrogen
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.