Learn More
CleanSpace™ Technology Bio Mask Exhalation Adaptor
Product Code. 17445382 Shop All CleanSpace Technology Products
Change view
Click to view available options
Bio Mask Adaptor; Material: Polyisoprene; Color: Blue; Design Type: Valve Cover; Dimensions (L x W x H): 18 x 6 x 5 cm; Disposable: Yes; For Use With (Equipment): CS3027
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17445382 Bio Mask Adaptor; Material: Polyisoprene; Color: Blue; Design Type: Valve Cover; Dimensions (L x W x H): 18 x 6 x 5 cm; Disposable: Yes; For Use With (Equipment): CS3027
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Product Code. 17445382 missing translation for 'mfr' CleanSpace™ Technology missing translation for 'supplierNo' CS3026
  • ONLINE EXCLUSIVE PRICE
    78.01€
    60.50€ / pezzi 5
    Risparmia 17.51€
    22% Sconto
Spedizione stimata: 18-12-2026
For use with the HALO system and is specifically designed for biocontainment units, laboratories, and pharmaceutical cleanrooms.
The BioHood System is compatible with the CleanSpace™ HALO Respirator. It is specifically designed for biocontainment units, laboratories and pharmaceutical cleanrooms, providing eye and head protection.
  • DuPont™ Tyvek™ material and seam technology
  • Light and durable
  • Blood and fluid resistant
  • Only PAPR hood design to completely cover the respirator when worn, minimizing device contamination
  • Can be easily removed while still wearing the respirator to protect healthcare professionals when doffing
  • Broad field of vision and unique design that secures the visor to eliminate slippage and movement
  • No belts, hoses, fogging or heat
  • Comfortable air flow with no breathing resistance
  • NIOSH and CE Mark approved
  • BioHood Parts: The BioHood (CS3024), Bio HEPA P3/T3 (CS3025), Bio Mask Exhalation Adaptor (CS3026) - reusable and attaches to CleanSpace™ HALO Face Mask (face mask sold separately)
Specifications
Color Blue
Disposable Single-use
Material Polyisoprene
Product Type Exhalation Adapter
For Use With (Equipment) CS3027
Model CS3026
Quantity 5/Pk.
Dimensions (L x W x H) 18 x 6 x 5 cm