Learn More
Ansell BIOCLEAN™ Permaflow Sterile Cleanroon Pen S-BPFP-RD1
Product Code. 30040336
Change view
Click to view available options
Color:
Red
Unit Size:
Pack of 200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30040336 Red Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30040336 Supplier Ansell Supplier No. SBPFPRD1
Spedizione stimata: 23-06-2027
Ballpoint pens for alcohol resistant labelling are processed to ensure ISO Class 4 and EU GMP Grade A compatibility, fade and water resistant, quick drying, and non-toxic.
  • BioClean Permaflow Cleanroom Pens are sterile with fade and water resistant super permanent ink.
  • With alcohol resistant labeling for easy cleaning within a critical environment, they are ISO Class 4 and EU GMP Grade A compatible.
TRUSTED_SUSTAINABILITY
Specifications
Tip Style Fine
Product Type Permanent Pen
Color Red