Learn More
Unity Lab Services™ Biosafety Cabinet Requalification Service
Product Code. 30178678
Change view
Click to view available options
For Use With (Equipment):
Biosafety Cabinet
Unit Size:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
30178678 Biosafety Cabinet 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30178678 Supplier Unity Lab Services™ Supplier No. RQBSC
 Restricted Item
Spedizione stimata: 15-12-2026
Requalification Services for Biosafety Cabinet (documentation included)
Requalification (RQ): Documented verification that equipment (as installed) is operating as intended according to manufacturers specifications.
  • Verifies important equipment functions and helps ensure that the equipment operates as expected by the manufacturer and conforms to standards and requirements.
  • Key tests offered based on relevant certification Standard (NSF49/en 12469/ DIN12480)
    • Few examples: Filter leakage, DEHS, Air velocity, Electrical safety, Air-flow smoke pattern, KI-DISCUS.
  • Please note that installation and operational qualification, ideally along with preventive maintenance and calibration, should be completed on lab equipment before requalification services can be performed.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Biosafety Cabinet
Product Type Requalification Service