Learn More
Welch Ilmvac™ Biovac 106 Aspiration System
Codice prodotto. 11856493 Sfoglia Tutto Welch Ilmvac Prodotti
Cambia vista
Click to view available options
Portata:
15 L/min.
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Portata unitSize
11856493 15 L/min. Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 11856493 Fornitore Welch Ilmvac™ N. del fornitore 112037
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Use this aspirator for aspiration and disposal of liquids in biochemistry, microbiology and cell culture applications. Welch Ilmvac™ Biovac 106 Aspiration Systems are idea for precise aspiration of chemical, biological and medical liquids.
  • Aspiration and filtration system
  • Includes handheld HandVac pipettor
  • Autoclavable collection receiver
  • Automatic flow stop and in-line hydrophobic filter
  • Available with 2 liter glass collection receiver
TRUSTED_SUSTAINABILITY
Specifica
Requisiti elettrici 115/230 V 50/60 Hz
Modello Biovac 106 Aspiration System
Peso (sistema metrico) 8 kg
Portata 15 L/min.
Tipo di pompa Aspiration System
Maximum Pressure <100 mbar