Learn More
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)barium(II), 99.99% (metals basis excluding Sr) Sr< 500ppm, Thermo Scientific™
Catalog Number p-7051493
Click to view available options
Quantità:
1 g
5 g
17594-47-7
C22H38BaO4
503.87
MFCD00058709
VJRLFEOHKCDHBT-UHFFFAOYSA-N
barium-dpm, ba tmhd 2, bis 2,2,6,6-tetramethyl-3,5-heptanedionato barium ii, barium 2,2,6,6-tetramethyl-3,5-heptanedionate tetramer, barium bis 2,2,6,6-tetramethyl-3,5-heptanedionate hydrate, bis 2,2,6,6-tetramethyl-3,5-heptanedionate barium ii, barium-thd, barium bis dipivaloylmethanate, barium ii 2,2,6,6-tetramethylheptanedionate, bis 2,2,6,6-tetramethyl-3,5 heptanedionato barium ii
17749137
barium(2+);(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate;hydrate
[Ba++].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C
17594-47-7
C22H38BaO4
503.87
MFCD00058709
VJRLFEOHKCDHBT-UHFFFAOYSA-N
barium-dpm, ba tmhd 2, bis 2,2,6,6-tetramethyl-3,5-heptanedionato barium ii, barium 2,2,6,6-tetramethyl-3,5-heptanedionate tetramer, barium bis 2,2,6,6-tetramethyl-3,5-heptanedionate hydrate, bis 2,2,6,6-tetramethyl-3,5-heptanedionate barium ii, barium-thd, barium bis dipivaloylmethanate, barium ii 2,2,6,6-tetramethylheptanedionate, bis 2,2,6,6-tetramethyl-3,5 heptanedionato barium ii
17749137
barium(2+);(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate;hydrate
[Ba++].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)barium(II)
Punto di fusione 200°C
Punti di ebollizione 225°C (0.3 mmHg)
Note percentuale saggio (metals basis excluding Sr) Sr < 500ppm
Quantità 1 g
Numero UN UN1564
Sensibilità Hygroscopic
Peso formulazione 503.85
Percent Purity 99.99%
Forma fisica Powder