Learn More
Bis(2-hydroxyethyl) Terephthalate, ~85%, TRC
Specifica
Specifica
| Materiale o nome chimico | Terephthalic Acid, Bis-2-hydroxyethyl Ester |
| CAS | 959-26-2 |
| Formula molecolare | C12 H14 O6 |
| Quantità | 50 g |
| Sinonimo | 1,4-Benzenedicarboxylic acid, bis(2-hydroxyethyl) ester (9CI), Ethylene glycol, terephthalate (2:1) (8CI), Terephthalic acid, bis(2-hydroxyethyl) ester (6CI, 8CI), BHET, BHET(J), BHET-N, Bis(2-hydroxyethyl) 1,4-benzenedicarboxylate, Bis(2-hydroxyethyl) te |
| InChI Formula | InChI=1S/C12H14O6/c13-5-7-17-11(15)9-1-2-10(4-3-9)12(16)18-8-6-14/h1-4,13-14H,5-8H2 |
| SMILES | OCCOC(=O)c1ccc(cc1)C(=O)OCCO |
| IUPAC Name | bis(2-hydroxyethyl) benzene-1,4-dicarboxylate |
| Molecular Weight (g/mol) | 254.236 |
| Peso formulazione | 254.079 |