missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcam Bis(3,5-dibromosalicyl) Fumarate, Hemoglobin Acylating Agent, 671.9 Da, 5 g
Descrizione
- Molecular weight: 671.9 Da
- Purity: >93%
- Potent hemoglobin acylating agent
- Cross-links beta chains of hemoglobin
- Acylating and anti-sickling agent
- Active in vitro and in vivo
- CAS number: 71337-53-6
Specifica
Specifica
| Tampone | 100% Bis(3,5-dibromosalicyl) fumarate |
| Content And Storage | Store at -20°C. |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Substrato | Hemoglobin |
| Target Molecule | Hemoglobin |
| Formulazione | 100% Bis(3,5-dibromosalicyl) fumarate |
| Quantità | 5 g |
| Da utilizzare con (applicazione) | Hemoglobin acylation, anti-sickling research |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?