missing translation for 'onlineSavingsMsg'
Learn More

Bis(3-fluorophenyl) disulfide, 97%, Thermo Scientific™

Catalog Number p-7047009
Click to view available options
Quantità:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
63930-17-6
C12H8F2S2
254.313
MFCD01318104
LFVFHTBHAIVNJQ-UHFFFAOYSA-N
bis 3-fluorophenyl disulfide, bis 3-fluorophenyl disulphide, 1,2-bis 3-fluorophenyl disulfane, 3,3'-difluoro diphenyl disulfide, 1-fluoro-3-3-fluorophenyl disulfanyl benzene, 3,3'-difluorodiphenyl disulfide, bis-3-fluorophenyl disulfide, disulfide, bis 3-fluorophenyl, di3-fluorophenyl disulfide
2737794
1-fluoro-3-[(3-fluorophenyl)disulfanyl]benzene
C1=CC(=CC(=C1)SSC2=CC=CC(=C2)F)F
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
63930-17-6
C12H8F2S2
254.313
MFCD01318104
LFVFHTBHAIVNJQ-UHFFFAOYSA-N
bis 3-fluorophenyl disulfide, bis 3-fluorophenyl disulphide, 1,2-bis 3-fluorophenyl disulfane, 3,3'-difluoro diphenyl disulfide, 1-fluoro-3-3-fluorophenyl disulfanyl benzene, 3,3'-difluorodiphenyl disulfide, bis-3-fluorophenyl disulfide, disulfide, bis 3-fluorophenyl, di3-fluorophenyl disulfide
2737794
1-fluoro-3-[(3-fluorophenyl)disulfanyl]benzene
C1=CC(=CC(=C1)SSC2=CC=CC(=C2)F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico Bis(3-fluorophenyl) disulfide
Densità 1.27
Punti di ebollizione 308°C
Punto d'infiammabilità >110°C (230°F)
Indice di rifrazione 1.6185
Quantità 1 g
Beilstein 2530329
Peso formulazione 254.31
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'