missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-hydroxyphenyl) sulfone, 99%, Thermo Scientific Chemicals

Catalog Number p-7023608
37.06 EUR valido fino al 2024-03-29
Usa il codice promo "21615" per avere il tuo prezzo promozionale.


Click to view available options
Quantità:
250 g
1000 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
80-09-1
C12H10O4S
250.27
MFCD00002350
VPWNQTHUCYMVMZ-UHFFFAOYSA-N
4,4'-sulfonyldiphenol, bisphenol s, bis 4-hydroxyphenyl sulfone, phenol, 4,4'-sulfonylbis, 4,4'-dihydroxydiphenyl sulfone, 4-hydroxyphenyl sulfone, bis p-hydroxyphenyl sulfone, 4,4'-sulfonylbisphenol, diphone c
6626
CHEBI:34372
4-(4-hydroxyphenyl)sulfonylphenol
OC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(O)C=C1
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
80-09-1
C12H10O4S
250.27
MFCD00002350
VPWNQTHUCYMVMZ-UHFFFAOYSA-N
4,4'-sulfonyldiphenol, bisphenol s, bis 4-hydroxyphenyl sulfone, phenol, 4,4'-sulfonylbis, 4,4'-dihydroxydiphenyl sulfone, 4-hydroxyphenyl sulfone, bis p-hydroxyphenyl sulfone, 4,4'-sulfonylbisphenol, diphone c
6626
CHEBI:34372
4-(4-hydroxyphenyl)sulfonylphenol
OC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(O)C=C1

It is a reactant used in epoxy reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a reactant used in epoxy reactions.

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico Bis(4-hydroxyphenyl) sulfone
Punto di fusione 245°C to 250°C
Densità 1.366
Quantità 250 g
Beilstein 2052954
Informazioni di solubilità Insoluble in water.
Peso formulazione 250.28
Percent Purity 99%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'