missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-methoxyphenyl) carbonate, 97%, Thermo Scientific™

Catalog Number 11402988
Click to view available options
Quantità:
10 g
50 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
5676-71-1
C15H14O5
274.272
MFCD00798557
PVBTWSPZTNYNHG-UHFFFAOYSA-N
bis 4-methoxyphenyl carbonate, 4-methoxyphenyl 4-methoxyphenoxy formate, acmc-1arso, 4-methoxyphenyl carbonate
3793869
bis(4-methoxyphenyl) carbonate
COC1=CC=C(C=C1)OC(=O)OC2=CC=C(C=C2)OC
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
5676-71-1
C15H14O5
274.272
MFCD00798557
PVBTWSPZTNYNHG-UHFFFAOYSA-N
bis 4-methoxyphenyl carbonate, 4-methoxyphenyl 4-methoxyphenoxy formate, acmc-1arso, 4-methoxyphenyl carbonate
3793869
bis(4-methoxyphenyl) carbonate
COC1=CC=C(C=C1)OC(=O)OC2=CC=C(C=C2)OC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico Bis(4-methoxyphenyl) carbonate
Punto di fusione 86°C to 88°C
Quantità 50 g
Peso formulazione 274.28
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'