Learn More
Bis(tetraethylammonium) bis(2-thioxo-1,3-dithiole-4,5-dithiolato)zincate, 97%
Product Code. 11323936
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11323936 250 mg 250mg
11313936 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11323936 Supplier Thermo Scientific Alfa Aesar Supplier No. L17046.MD
 Restricted Item
Spedizione stimata: 25-11-2026
Add to basket
CAS: 72022-68-5 | C22H40N2S10Zn | 718.556 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 72022-68-5
Molecular Formula C22H40N2S10Zn
Molecular Weight (g/mol) 718.556
MDL Number MFCD00623815
InChI Key TXRIVAQWJHZFOT-UHFFFAOYSA-J
Synonym bis tetraethylammonium bis 2-thioxo-1,3-dithiole-4,5-dithiolato zincate
PubChem CID 15762389
IUPAC Name zinc;2-sulfanylidene-1,3-dithiole-4,5-dithiolate;tetraethylazanium
SMILES CC[N+](CC)(CC)CC.CC[N+](CC)(CC)CC.C1(=C(SC(=S)S1)[S-])[S-].C1(=C(SC(=S)S1)[S-])[S-].[Zn+2]
Specifications
Melting Point 202°C to 206°C
Quantity 250 mg
Beilstein 4900651
Sensitivity Light sensitive
Formula Weight 718.6
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Bis(tetraethylammonium) bis(2-thioxo-1,3-dithiole-4,5-dithiolato)zincate