missing translation for 'onlineSavingsMsg'
Learn More

Bis(triphenylphosphoranylidene)ammonium chloride, 97% (dry wt.), water <3%, Thermo Scientific Chemicals

Catalog Number p-7046442
Click to view available options
Quantità:
5 g
25 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
21050-13-5
C36H30ClNP2
574.04
MFCD00151523
LVRCYPYRKNAAMX-UHFFFAOYSA-M
1,1,1-triphenyl-n-triphenylphosphoranylidene phosphoraniminium chloride, bis triphenylphosphoranylidene ammonium chloride, bis triphenylphosphoranylidene ammonium, triphenyl p,p,p-triphenylphosphine imidato-n phosphorus 1+ tetracarbonylcobaltate 1-, bis triphenylphosphine iminium chloride∼ppncl, ppn chloride, ppn cl, ksc491c1r, parent, triphenyl p,p,p-triphenylphosphine imidato phosphorus 1+ chloride
3036656
triphenyl-[(triphenyl-$l^{5}-phosphanylidene)amino]phosphanium;chloride
[Cl-].C1=CC=C(C=C1)P(=[N+]=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
21050-13-5
C36H30ClNP2
574.04
MFCD00151523
LVRCYPYRKNAAMX-UHFFFAOYSA-M
1,1,1-triphenyl-n-triphenylphosphoranylidene phosphoraniminium chloride, bis triphenylphosphoranylidene ammonium chloride, bis triphenylphosphoranylidene ammonium, triphenyl p,p,p-triphenylphosphine imidato-n phosphorus 1+ tetracarbonylcobaltate 1-, bis triphenylphosphine iminium chloride∼ppncl, ppn chloride, ppn cl, ksc491c1r, parent, triphenyl p,p,p-triphenylphosphine imidato phosphorus 1+ chloride
3036656
triphenyl-[(triphenyl-$l^{5}-phosphanylidene)amino]phosphanium;chloride
[Cl-].C1=CC=C(C=C1)P(=[N+]=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Co-catalyst used with a chiral Cobalt-salen complex for the asymmetric addition of carbon dioxide to propylene oxide. Also used in the preparation of 5-Hydroxy-clethodim Sulfoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Co-catalyst used with a chiral Cobalt-salen complex for the asymmetric addition of carbon dioxide to propylene oxide. Also used in the preparation of 5-Hydroxy-clethodim Sulfoxide.

Solubility
Moderately soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico Bis(triphenylphosphoranylidene)ammonium chloride
Punto di fusione 274°C to 276°C
Note percentuale saggio (dry wt.), water <3%
Quantità 5 g
Beilstein 4097979
Informazioni di solubilità Moderately soluble in water.
Peso formulazione 574.05
Percent Purity 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'