Learn More
Bithionol, TRC
Specifica
Specifica
| Materiale o nome chimico | Bithionol |
| CAS | 97-18-7 |
| Formula molecolare | C12 H6 Cl4 O2 S |
| Quantità | 25 g |
| Sinonimo | 2,2'-Thiobis[4,6-dichlorophenol], 2,2-Thiobis(4,6-dichlorophenol), 2,2'-Dihydroxy-3,3',5,5'-tetrachlorodiphenyl sulfide, 2-Hydroxy-3,5-dichlorophenyl sulphide, Actamer, Bidiphen, Bis(2-hydroxy-3,5-dichlorophenyl) sulfide, Bisoxyphen, Bithin, Bithionol, Bitin, Bitionol, CP 3438, D 26, Lorothidol, Lorothiodol, NSC 47129, NSC 9872, Neopellis, Nobacter, TKhSD, Vancide BL, XL 7 |
| InChI Formula | InChI=1S/C12H6Cl4O2S/c13-5-1-7(15)11(17)9(3-5)19-10-4-6(14)2-8(16)12(10)18/h1-4,17-18H |
| SMILES | Oc1c(Cl)cc(Cl)cc1Sc2cc(Cl)cc(Cl)c2O |
| IUPAC Name | 2,4-dichloro-6-(3,5-dichloro-2-hydroxyphenyl)sulfanylphenol |
| Molecular Weight (g/mol) | 356.05 |
| Peso formulazione | 353.8843 |