missing translation for 'onlineSavingsMsg'
Learn More

Bithionol, TRC

Codice prodotto. 30338753
Cambia vista
Click to view available options
Quantità:
10 g
Dimensione della confezione:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30338753 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 30338753 Fornitore Toronto Research Chemicals N. del fornitore TRCB59136310G

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Materiale o nome chimico Bithionol
CAS 97-18-7
Formula molecolare C12 H6 Cl4 O2 S
Quantità 10 g
Sinonimo 2,2'-Thiobis[4,6-dichlorophenol], 2,2-Thiobis(4,6-dichlorophenol), 2,2'-Dihydroxy-3,3',5,5'-tetrachlorodiphenyl sulfide, 2-Hydroxy-3,5-dichlorophenyl sulphide, Actamer, Bidiphen, Bis(2-hydroxy-3,5-dichlorophenyl) sulfide, Bisoxyphen, Bithin, Bithionol, Bitin, Bitionol, CP 3438, D 26, Lorothidol, Lorothiodol, NSC 47129, NSC 9872, Neopellis, Nobacter, TKhSD, Vancide BL, XL 7
InChI Formula InChI=1S/C12H6Cl4O2S/c13-5-1-7(15)11(17)9(3-5)19-10-4-6(14)2-8(16)12(10)18/h1-4,17-18H
SMILES Oc1c(Cl)cc(Cl)cc1Sc2cc(Cl)cc(Cl)c2O
IUPAC Name 2,4-dichloro-6-(3,5-dichloro-2-hydroxyphenyl)sulfanylphenol
Molecular Weight (g/mol) 356.05
Peso formulazione 353.8843
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.