Learn More
Uvex Black/Silver S2 69798 Motion Style Boot - Width 11
Uvex Motion Style safety shoes are lightweight with optimizes wearer comfort using Climazone climate management
Marca:  Uvex 6979850
Dettagli aggiuntivi : Peso : 1.96200kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17244331
  • 133.00€ / 1 paio
Spedizione stimata: 08-11-2022
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
Uvex Motion Style Lace-Up Boot 6979 S2 SRC
  • Extremely light, fashionable shoe
  • Breathable, water-resistant high-tech microvelour
  • Ultra breathable distance mesh lining
  • Made from synthetic textiles, suitable for people allergic to chrome
  • Uvex Climazone technology for optimum comfort
  • Steel toecap and stabilizing Uvex anti-twist heel cap for greater stability and protection from ankle twisting
  • Soft padded collar
  • Comfortable removable insole, antistatic with moisture control
  • Uvex 3D Hydroflex™ foam insole for exceptional shock absorption in heel and under front of foot
  • Fulfills ESD requirements, resistance to ground Kleiner 35 megohms
  • PUR outsole
  • Article No. 6979.8
  • Standard EN ISO 20345:2011 S2 SRC
  • Sizes: 36 to 50
Specifica
Specifica
Silver, Black
50
Steel
EN ISO 20345:2011
Standard width
Yes
S2
Microvelour
Dual-density polyurethane (PU/PU)
No
Unisex
Uvex Motion Classic
No
SRC