Learn More
Brady™ Breaker Blocker Components - Blocking Bar Holder
Product Code. 15683487 Shop All Brady Products
Change view
Click to view available options
Color:
Yellow
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15683487 Yellow Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15683487 Supplier Brady™ Supplier No. 051265
  • ONLINE EXCLUSIVE PRICE
    42.95€
    39.46€ / pezzi 5
    Risparmia 3.49€
    8% Sconto
In stock
For use with Bradys 480/600V Breaker Blocker
Specifications
Product Type Blocking Bar Holder
Alternate Part Number BREAKER BLOCKER ARM HOLDER 5 PACK
Color Yellow
Description Breaker Blocker Components - Blocking Bar Holder
Quantity 1 Ea.