Learn More
BD Safety-Lok™ Blood Collection Set
Product Code. 15706238
Change view
Click to view available options
Sterility:
Sterile
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Unit Size
15706238 Sterile Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15706238 Supplier BD Supplier No. 367282
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
 Restricted Item
Spedizione stimata: 13-01-2027
Use for blood collection. BD Vacutainer™ Safety-Lok™ Blood Collection Set (50 sets) comes with safety re-shielding cover and Luer adapter.
  • Includes safety re-shielding cover and Luer adapter,
  • 21 G ¾ in., 18 cm tubing, green
  • Recommended for adults
  • One case consists fo 4 x 50 Blood Collection Sets
  • One Set is necessary for a pair of blood culture bottles
TRUSTED_SUSTAINABILITY
Specifications
Product Type Blood Collection Set
Sterility Sterile