Learn More
Vitlab™ Blue Raised Scale Graduated Beaker
Product Code. 11782729
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
3000 mL
5000 mL
Unit Size:
Each
Pack of 12
Pack of 24
Pack of 6
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11782729 5000 mL Pack of 6
10521833 50 mL Pack of 24
10757981 100 mL Each
10689622 100 mL Pack of 24
10709242 250 mL Pack of 12
10605173 3000 mL Pack of 6
10602821 5000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11782729 Supplier Vitlab™ Supplier No. 445081
  • ONLINE EXCLUSIVE PRICE
    130.00€
    128.10€ / pezzi 6
    Risparmia 1.90€
    1% Sconto
In stock
Add to basket
Edge
Add to basket
Use these polypropylene beakers for a variety of general laboratory applications. VITLAB™ Blue Raised Scale Graduated Beakers feature a stable handle easy-grip handle and an easily readable, embossed blue scale.
  • Highly transparent polypropylene beaker
  • To preserve markings, do not clean at temperatures exceeding 60°C
TRUSTED_SUSTAINABILITY
Specifications
Color Transparent
Diameter (Metric) Outer 210 mm
Capacity (Metric) 5000 mL
Material Polypropylene
Height (Metric) 270 mm
Quantity 6 Pk.
Type Beaker
Autoclavable Autoclavable
Graduated Yes
Certifications/Compliance DIN EN 285
Scale Blue Raised Scale