missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BMP-6 Polyclonal Antibody

Descrizione
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
BMP6 is a member of bone morphogenetic proteins (BMPs) are a family of secreted signaling molecules that can induce ectopic bone growth. Many BMPs are part of the transforming growth factor-beta (TGFB) superfamily. BMPs were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. Based on its expression early in embryogenesis, Bmp6 has a proposed role in early development. In addition, the fact that BMP6 is closely related to BMP5 and BMP7 has lead to speculation of possible bone inductive activity.
Specifica
Specifica
| Antigene | BMP-6 |
| Applicazioni | Immunohistochemistry (Paraffin), Western Blot |
| Classificazione | Polyclonal |
| Concentrazione | 1 mg/mL |
| Coniugato | Unconjugated |
| Formulazione | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | BMP6 |
| N. accesso geni | P20722, P22004, Q04906 |
| Alias gene | BMP; BMP 6; BMP6; BMP-6; Bone morphogenetic protein; Bone morphogenetic protein 6; D13Wsu115e; vegetal related growth factor (TGFB-related); vegetal-related (TGFB related) cytokine; VG-1-R; VG-1-related protein; Vg1-related sequence; Vgr; Vgr1; VGR-1 |
| Simboli geni | BMP6 |
| Vedi altri risultati |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?