missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BMP4 Recombinant Rabbit Monoclonal Antibody (7Q2U7)

Descrizione
Immunogen sequence: RRHSLYVDFS DVGWNDWIVA PPGYQAFYCH GDCPFPLADH LNSTNHAIVQ TLVNSVNSSI PKACCVPTEL SAISMLYLDE YDKVVLKNYQ EMVVEGCGCR.
The protein encoded by this gene is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. This particular family member plays an important role in the onset of endochondral bone formation in humans, and a reduction in expression has been associated with a variety of bone diseases, including the heritable disorder Fibrodysplasia Ossificans Progressiva. Alternative splicing in the 5' untranslated region of this gene has been described and three variants are described, all encoding an identical protein.
Specifica
Specifica
| Antigene | BMP4 |
| Applicazioni | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classificazione | Recombinant Monoclonal |
| Clone | 7Q2U7 |
| Concentrazione | 1 mg/mL |
| Coniugato | Unconjugated |
| Formulazione | PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | BMP4 |
| N. accesso geni | P12644 |
| Alias gene | BMP; BMP2B; BMP-2B; Bmp2b1; Bmp2b-1; Bmp4; Bmp-4; BOMPR4A; Bone morphogenetic protein; Bone morphogenetic protein 2B; bone morphogenetic protein 4; bone morphogenetic protein 4 preproprotein; DVR4; Dvr-4; MCOPS6; OFC11; ZYME |
| Vedi altri risultati |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?