Learn More
Brady™ BMP41 Label Printer for EU
Descrizione
The BMP41 is engineered with long-lasting material quality, and can print durable, true-sized 25 mm wide identification to maximize the amount of data on your label.
- Prints up to 250 labels per day for higher volume printing
- Quick to change supplies; just pop in the needed cartridge
- Large LCD backlit display provides clear visibility
- Drop-tested printer with rubber corner bumpers provide optimal protection and security from falls and wear and tear conditions
- Smart Cell Technology means your label format is automatically set up with no calibration
- Long-life rechargeable battery allows printing on the go and decreases need for frequent charging
- Includes Brady™ Workstation Basic Design Software Suite
Specifica
Specifica
| Tipo di display | Monocolor LCD |
| Print Technology | Trasferimento termico |
| Larghezza (sistema imperiale) | 6 in. |
| Larghezza (sistema metrico) | 152 mm |
| Altezza (sistema imperiale) | 11.5 in. |
| Altezza (sistema metrico) | 292 mm |
| Profondità (sistema imperiale) | 3.5 in. |
| Profondità (sistema metrico) | 89 mm |
| Autoclavabile | Not Autoclavable |
| Sterilità | Non-sterile |
| Vedi altri risultati |