missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BMS 303141

Codice prodotto. 30001590
Cambia vista
Click to view available options
Quantità:
10 mg
50 mg
Dimensione della confezione:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30001590 10 mg 10mg
30001546 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 30001590 Fornitore Tocris Bioscience™ N. del fornitore 4609/10

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

ATP citrate lyase inhibitor; orally bioavailable

ATP citrate lyase (ACL) inhibitor (IC50 = 0.13 μM for human recombinant ACL); blocks lipid synthesis (IC50 = 8 μM in HepG2 cells). Displays no cytotoxicity up to a concentration of 50 μM. Lowers plasma glucose and triglycerides in a mouse model of hyperlipidemia. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 943962-47-8
Formula molecolare C19H15Cl2NO4S

Specifica

Materiale o nome chimico BMS 303141
Quantità 10 mg
Peso formulazione 424.3
Purezza 0.98
Peso molecolare 424.29
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.