missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boc-ON, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantità:
5 g
25 g
100 g
Descrizione
Widely used for protection of amino groups in peptide synthesis and multi-step organic synthesis of small molecules.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Widely used for protection of amino groups in peptide synthesis and multi-step organic synthesis of small molecules.
Solubility
Soluble in ethyl acetate, ether and benzene. Insoluble in water.
Notes
Moisture Sensitive, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
Widely used for protection of amino groups in peptide synthesis and multi-step organic synthesis of small molecules.
Solubility
Soluble in ethyl acetate, ether and benzene. Insoluble in water.
Notes
Moisture Sensitive, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
Specifica
Materiale o nome chimico | Boc-ON |
Punto di fusione | 86°C to 89°C |
Quantità | 5 g |
Numero UN | UN3439 |
Beilstein | 2217296 |
Sensibilità | Moisture sensitive |
Informazioni di solubilità | Soluble in ethyl acetate,ether and benzene. Insoluble in water. |
Peso formulazione | 246.27 |
Percent Purity | ≥98% |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'