Learn More
Bohlender™ BOLA™ Flexible Distributors
Descrizione
- Screw cap blue made of PP
- For bottle thread GLS 80
- Distributor body with hose connectors made of PP
- Bent hose connectors on the upper side, straight hose connectors on the bottom side
- Restricted chemical resistance
Specifica
Specifica
| Colore | Blue |
| Diametro (sistema imperiale) interno | 0.24 in. |
| Diametro (sistema metrico) interno | 6 mm |
| Include | 3 Hose Connectors |
| Materiale | Polypropylene |
| Temperatura max (sistema metrico) | 110°C |
| Temperatura max (sistema imperiale) | 230°F |
| Dimensioni filettatura (sistema metrico) | GLS 80 |
| Diameter (Metric) Outer Tubing | 9 mm, 12 mm |
| Product Type | Distributor Cap |