missing translation for 'onlineSavingsMsg'
Learn More

Boldenone Propionate, TRC

Codice prodotto. 30331758
Change view
Click to view available options
Quantità:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità unitSize
30331758 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331758 Supplier Toronto Research Chemicals Supplier No. TRCB675160500MG

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Materiale o nome chimico 3-Oxoandrosta-1,4-dien-17beta-yl Propanoate
CAS 977-32-2
Note sul grado di purezza HPLC
Formula molecolare C22 H30 O3
Quantità 500 mg
Sinonimo 3-Oxoandrosta-1,4-dien-17β-yl propanoate, Androsta-1,4-dien-3-one, 17-(1-oxopropoxy)-, (17β)-, Androsta-1,4-dien-3-one, 17β-hydroxy-, propionate (6CI,7CI,8CI), (17β)-17-(1-Oxopropoxy)androsta-1,4-dien-3-one, Boldenone propionate
InChI Formula InChI=1S/C22H30O3/c1-4-20(24)25-19-8-7-17-16-6-5-14-13-15(23)9-11-21(14,2)18(16)10-12-22(17,19)3/h9,11,13,16-19H,4-8,10,12H2,1-3H3/t16-,17-,18-,19-,21-,22-/m0/s1
SMILES CCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-17-yl] propanoate
Molecular Weight (g/mol) 342.47
Peso formulazione 342.2195
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.